Cy-vBRIDP® ≥90.0% (by HPLC)
Supplier: TCI America
C2981-1G
C2981-200MG
TCC2981-1GEA
171.9
CAD
TCC2981-1G
TCC2981-200MG
Cy-vBRIDP® ≥90.0% (by HPLC)
Dicyclohexyl(1,1-diphenylprop-1-en-2-yl)phosphine
CAS Number: 384842-24-4
MDL Number: MFCD10565627
Molecular Formula: C27H35P
Molecular Weight: 390.55
Purity/Analysis Method:
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: Dicyclohexyl(1,1-diphenyl-1-propen-2-yl)phosphine
Formula:
C₂₇H₃₅P Melting Pt: 130 °C |
MDL Number:
MFCD10565627 CAS Number: 384842-24-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...