7-Chloro-1-tetralone ≥95.0%

Supplier: TCI America

Synonyms: 7-Chloro-3,4-dihydronaphthalen-1(2H)-one, 7-Chloro-3,4-dihydro-1(2H)-naphthalenone

C2976-1G C2976-5G
TCC2976-1GEA 66.04 CAD
TCC2976-1G TCC2976-5G
7-Chloro-1-tetralone ≥95.0%
7-Chloro-1-tetralone

CAS Number: 26673-32-5
MDL Number: MFCD01928386
Molecular Formula: C10H9ClO
Molecular Weight: 180.63
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 95
Lambda max.: 305 nm (MeOH)


Other Product Names / Synonyms: 7-Chloro-3,4-dihydro-1(2H)-naphthalenone

Formula: C₁₀H₉ClO
MW: 180.63 g/mol
Boiling Pt: 115 °C
Melting Pt: 95 °C
MDL Number: MFCD01928386
CAS Number: 26673-32-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=135100a414c903bbd6c4e53f58eb7c05aee941fa&vid=135100a414c903bbd6c4e53f58eb7c05aee941fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR