5'-Chloro-2'-hydroxy-3'-nitroacetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-Chlor-3-ethanoyl-4-hydroxy-5-nitrobenzene, 1-(5-Chloro-2-hydroxy-3-nitrophenyl)ethanone

C2973-25G C2973-5G
TCC2973-25GEA 142.2 CAD
TCC2973-25G TCC2973-5G
5'-Chloro-2'-hydroxy-3'-nitroacetophenone ≥98.0%
5'-Chloro-2'-hydroxy-3'-nitroacetophenone

CAS Number: 84942-40-5
MDL Number: MFCD00191921
Molecular Formula: C8H6ClNO4
Molecular Weight: 215.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 133

Formula: C₈H₆ClNO₄
Melting Pt: 133 °C
MDL Number: MFCD00191921
CAS Number: 84942-40-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7af9488d3d4ea00f3af80b8a1f216ef2d6c3d694&vid=7af9488d3d4ea00f3af80b8a1f216ef2d6c3d694&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR