4,4'-Carbonyldimorpholine ≥98.0%
Supplier: TCI America
Synonyms:
Dimorpholinomethanone
C2962-1G
C2962-5G
TCC2962-1GEA
347.6
CAD
TCC2962-1G
TCC2962-5G
4,4'-Carbonyldimorpholine ≥98.0%
4,4'-Carbonyldimorpholine
CAS Number: 38952-62-4
MDL Number: MFCD06200603
Molecular Formula: C9H16N2O3
Molecular Weight: 200.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 144
Formula:
C₉H₁₆N₂O₃ MW: 200.24 g/mol Melting Pt: 144 °C |
MDL Number:
MFCD06200603 CAS Number: 38952-62-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...