2-Chloro-5-methylpyrimidine ≥98.0% (by GC)

Supplier: TCI America

C2957-1G C2957-5G
TCC2957-1GEA 64.78 CAD
TCC2957-1G TCC2957-5G
2-Chloro-5-methylpyrimidine ≥98.0% (by GC)
2-Chloro-5-methylpyrimidine

CAS Number: 22536-61-4
MDL Number: MFCD09260903
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 94

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Melting Pt: 94 °C
MDL Number: MFCD09260903
CAS Number: 22536-61-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f17526187ae77e0ae036ea9c80501892cf539d3e&vid=f17526187ae77e0ae036ea9c80501892cf539d3e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR