3-Chlorophenethylamine ≥98.0%

Supplier: TCI America

Synonyms: 2-(3-Chlorophenyl)ethylamine

C2955-25G C2955-5G
TCC2955-25GEA 259.12 CAD
TCC2955-25G TCC2955-5G
3-Chlorophenethylamine ≥98.0%
3-Chlorophenethylamine

CAS Number: 13078-79-0
MDL Number: MFCD00047957
Molecular Formula: C8H10ClN
Molecular Weight: 155.63
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 113
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 3-Chlorophenethylamine

Formula: C₈H₁₀ClN
Boiling Pt: 145 °C
Density: 1.13 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00047957
CAS Number: 13078-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87e0f6b0b6e21b91cd49f7b4a65a6869ac6c79c0&vid=87e0f6b0b6e21b91cd49f7b4a65a6869ac6c79c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR