2-Chloro-6-methylquinoline-3-carbaldehyde ≥96.0%

Supplier: TCI America

Synonyms: 2-Chloro-6-methylquinoline-3-carboxaldehyde, 2-Chloro-3-formyl-6-methylquinoline

C2953-25G C2953-5G
TCC2953-25GEA 467.68 CAD
TCC2953-25G TCC2953-5G
2-Chloro-6-methylquinoline-3-carbaldehyde ≥96.0%
2-Chloro-6-methylquinoline-3-carbaldehyde

CAS Number: 73568-27-1
MDL Number: MFCD01912773
Molecular Formula: C11H8ClNO
Molecular Weight: 205.64
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 124


Other Product Names / Synonyms: 2-Chloro-3-formyl-6-methylquinoline

Formula: C₁₁H₈ClNO
MW: 205.6400 g/mol
Melting Pt: 124 °C
MDL Number: MFCD01912773
CAS Number: 73568-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bcb8d09f181619db6f26b658c90d74af8335621&vid=9bcb8d09f181619db6f26b658c90d74af8335621&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR