(4-Chloro-2-pyridinyl)methanol ≥98.0%
Supplier: TCI America
C2950-1G
C2950-5G
TCC2950-1GEA
54.51
CAD
TCC2950-1G
TCC2950-5G
(4-Chloro-2-pyridinyl)methanol ≥98.0%
(4-Chloro-2-pyridinyl)methanol
CAS Number: 63071-10-3
MDL Number: MFCD07437885
Molecular Formula: C6H6ClNO
Molecular Weight: 143.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 66
Melting point (°C): 45
Other Product Names / Synonyms: 4-Chloro-2-(hydroxymethyl)pyridine
Formula:
C₆H₆ClNO Boiling Pt: 66 °C Melting Pt: 45 °C |
MDL Number:
MFCD07437885 CAS Number: 63071-10-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...