4-Chloro-6,7-dimethoxyquinoline ≥98.0%

Supplier: TCI America

C2938-25G C2938-5G
TCC2938-25GEA 568.64 CAD
TCC2938-25G TCC2938-5G
4-Chloro-6,7-dimethoxyquinoline ≥98.0%
4-Chloro-6,7-dimethoxyquinoline

CAS Number: 35654-56-9
MDL Number: MFCD07778437
Molecular Formula: C11H10ClNO2
Molecular Weight: 223.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 134

Formula: C₁₁H₁₀ClNO₂
MW: 223.6556 g/mol
Melting Pt: 134 °C
MDL Number: MFCD07778437
CAS Number: 35654-56-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca27979a514210741a2d9e10f46745732f27eec6&vid=ca27979a514210741a2d9e10f46745732f27eec6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR