6-Chloro-1-tetralone ≥96.0%

Supplier: TCI America

Synonyms: 6-Chloro-3,4-dihydronaphthalen-1(2H)-one, 6-Chloro-3,4-dihydro-1(2H)-naphthalenone

C2917-1G C2917-5G
TCC2917-1GEA 198.29 CAD
TCC2917-1G TCC2917-5G
6-Chloro-1-tetralone ≥96.0%
6-Chloro-1-tetralone

CAS Number: 26673-31-4
MDL Number: MFCD04114377
Molecular Formula: C10H9ClO
Molecular Weight: 180.63
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 150
Freezing point (°C): 28


Other Product Names / Synonyms: 6-Chloro-3,4-dihydro-1(2H)-naphthalenone

Formula: C₁₀H₉ClO
MW: 180.63 g/mol
Boiling Pt: 150 °C
MDL Number: MFCD04114377
CAS Number: 26673-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ee7f65487ce3ded562474fdd936e6c8f5d25f2b&vid=7ee7f65487ce3ded562474fdd936e6c8f5d25f2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR