3-Chloro-5-fluorophenylboronic acid 97.0-112.0% (contains varying amounts of Anhydride)

Supplier: TCI America

C2914-1G C2914-5G
TCC2914-1GEA 60.04 CAD
TCC2914-1G TCC2914-5G
3-Chloro-5-fluorophenylboronic acid 97.0-112.0% (contains varying amounts of Anhydride)
3-Chloro-5-fluorophenylboronic acid

CAS Number: 328956-61-2
MDL Number: MFCD06801741
Molecular Formula: C6H5BClFO2
Molecular Weight: 174.36
Form: Crystal
Color: White


Other Product Names / Synonyms: 3-Chloro-5-fluorobenzeneboronic Acid

Formula: C₆H₅BClFO₂
MDL Number: MFCD06801741
CAS Number: 328956-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e77f9404b030b27e4d124da1e029526b1f80e781&vid=e77f9404b030b27e4d124da1e029526b1f80e781&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR