4-Cyano-4'-propoxybiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4'-Propoxy-[1,1'-biphenyl]-4-carbonitrile

C2911-1G C2911-5G
TCC2911-1GEA 60.04 CAD
TCC2911-1G TCC2911-5G
4-Cyano-4'-propoxybiphenyl ≥98.0%
4-Cyano-4'-propoxybiphenyl

CAS Number: 52709-86-1
MDL Number: MFCD00183206
Molecular Formula: C16H15NO
Molecular Weight: 237.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73
Lambda max.: 294 nm (EtOH)


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4'-Propoxy-4-biphenylcarbonitrile, 3OCB

Formula: C₁₆H₁₅NO
MW: 237.3 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00183206
CAS Number: 52709-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d75724ef09dbe421027fb60497a8a9a09d3e317&vid=5d75724ef09dbe421027fb60497a8a9a09d3e317&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR