4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol
C2909-25G
C2909-5G
TCC2909-25GEA
1004.88
CAD
TCC2909-25G
TCC2909-5G
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidine ≥98.0% (by GC, titration analysis)
4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-hydroxypiperidine
CAS Number: 21928-50-7
MDL Number: MFCD00066999
Molecular Formula: C12H13ClF3NO
Molecular Weight: 279.69
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 140
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-[4-Chloro-3-(trifluoromethyl)phenyl]-4-piperidinol
Formula:
C₁₂H₁₃ClF₃NO MW: 279.69 g/mol Melting Pt: 140 °C |
MDL Number:
MFCD00066999 CAS Number: 21928-50-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...