4-Chloro-3-nitrobenzophenone ≥98.0%
Supplier: TCI America
C2906-25G
C2906-5G
TCC2906-25GEA
139.04
CAD
TCC2906-25G
TCC2906-5G
4-Chloro-3-nitrobenzophenone ≥98.0%
4-Chloro-3-nitrobenzophenone
CAS Number: 56107-02-9
MDL Number: MFCD00007081
Molecular Formula: C13H8ClNO3
Molecular Weight: 261.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 235
Melting point (°C): 106
Lambda max.: 260 nm
Formula:
C₁₃H₈ClNO₃ MW: 261.66 g/mol Boiling Pt: 235 °C Melting Pt: 106 °C |
MDL Number:
MFCD00007081 CAS Number: 56107-02-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...