3-Chloro-1-phenylpropan-1-ol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Chloro-1-phenyl-1-propanol, 3-Chloro-1-phenyl-propanol, alpha-(2-Chloroethyl)benzyl alcohol

C2904-25G C2904-5G
TCC2904-25GEA 619.36 CAD
TCC2904-25G TCC2904-5G
3-Chloro-1-phenylpropan-1-ol ≥98.0% (by GC)
3-Chloro-1-phenylpropan-1-ol

CAS Number: 18776-12-0
MDL Number: MFCD09878800
Molecular Formula: C9H11ClO
Molecular Weight: 170.64
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 116
Melting point (°C): 32


Other Product Names / Synonyms: alpha-(2-Chloroethyl)benzyl Alcohol

Formula: C9H11ClO
MW: 170.6400 g/mol
Boiling Pt: 116 °C
Melting Pt: 32 °C
MDL Number: MFCD09878800
CAS Number: 18776-12-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=041367469924b268e3189c357886e70e6976ef92&vid=041367469924b268e3189c357886e70e6976ef92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR