(3-Chloro-4-methoxyphenyl)methanamine hydrochloride ≥97.0%

Supplier: TCI America

Synonyms: 3-Chloro-4-methoxybenzylamine hydrochloride

C2903-1G C2903-5G
TCC2903-1GEA 26.86 CAD
TCC2903-1G TCC2903-5G
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride ≥97.0%
(3-Chloro-4-methoxyphenyl)methanamine hydrochloride

CAS Number: 41965-95-1
MDL Number: MFCD01211460
Molecular Formula: C8H10ClNO
Molecular Weight: 208.08
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 252

Formula: C₈H₁₁Cl₂NO
MW: 208.09 g/mol
Melting Pt: 252 °C
MDL Number: MFCD01211460
CAS Number: 41965-95-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b4cdd5b09b096a2125463b4525a9ff2e120314b&vid=2b4cdd5b09b096a2125463b4525a9ff2e120314b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR