(3-Chlorophenyl)acetone ≥95.0%

Supplier: TCI America

Synonyms: 1-(3-Chlorophenyl)propan-2-one

C2895-25G C2895-5G
TCC2895-25GEA 583.02 CAD
TCC2895-25G TCC2895-5G
(3-Chlorophenyl)acetone ≥95.0%
(3-Chlorophenyl)acetone

CAS Number: 14123-60-5
MDL Number: MFCD00082872
Molecular Formula: C9H9ClO
Molecular Weight: 168.62
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: 1-(3-Chlorophenyl)-2-propanone

Formula: C₉H₉ClO
Boiling Pt: 99 °C
Density: 1.16 (20 °C)
MDL Number: MFCD00082872
CAS Number: 14123-60-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c10f79a5d1f2f632b70b1cdda8398f6017e4f5a5&vid=c10f79a5d1f2f632b70b1cdda8398f6017e4f5a5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR