1-Cyclopentenylboronic acid 93.0-120.0% (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Cyclopent-1-en-1-ylboronic acid

C2893-1G C2893-5G
TCC2893-1GEA 118.5 CAD
TCC2893-1G TCC2893-5G
1-Cyclopentenylboronic acid 93.0-120.0% (contains varying amounts of Anhydride)
1-Cyclopentenylboronic acid

CAS Number: 850036-28-1
MDL Number: MFCD02179496
Molecular Formula: C5H9BO2
Molecular Weight: 111.94
Form: Crystal
Color: White
Melting point (°C): 140


Other Product Names / Synonyms: Cyclopentene-1-boronic Acid

Formula: C₅H₉BO₂
MW: 111.94 g/mol
Melting Pt: 140 °C
MDL Number: MFCD02179496
CAS Number: 850036-28-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d600fbf436b36a8bd02e3a22c41854b443413341&vid=d600fbf436b36a8bd02e3a22c41854b443413341&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR