4-Cyano-4'-ethylbiphenyl ≥98.0%
Supplier: TCI America
Synonyms:
4'-Ethyl-[1,1'-biphenyl]-4-carbonitrile
C2892-25G
C2892-5G
TCC2892-25GEA
443.03
CAD
TCC2892-25G
TCC2892-5G
4-Cyano-4'-ethylbiphenyl ≥98.0%
4-Cyano-4'-ethylbiphenyl
CAS Number: 58743-75-2
MDL Number: MFCD00799420
Molecular Formula: C15H13N
Molecular Weight: 207.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 205
Melting point (°C): 77
Other Product Names / Synonyms: 4'-Ethyl-4-biphenylcarbonitrile, 2CB
Formula:
C₁₅H₁₃N MW: 207.27 g/mol Boiling Pt: 205 °C Melting Pt: 77 °C |
MDL Number:
MFCD00799420 CAS Number: 58743-75-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...