4-Chloro-1H-pyrazole ≥98.0%

Supplier: TCI America

C2888-25G C2888-5G
TCC2888-25GEA 176.96 CAD
TCC2888-25G TCC2888-5G
4-Chloro-1H-pyrazole ≥98.0%
4-Chloro-1H-pyrazole

CAS Number: 15878-00-9
MDL Number: MFCD00159634
Molecular Formula: C3H3ClN2
Molecular Weight: 102.52
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 77
Lambda max.: 224 nm

Formula: C₃H₃ClN₂
MW: 102.5200 g/mol
Boiling Pt: 220 °C
Melting Pt: 77 °C
MDL Number: MFCD00159634
CAS Number: 15878-00-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5308a3f996c33257e7716744c8524b48a697717&vid=f5308a3f996c33257e7716744c8524b48a697717&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR