(2-Carboxyethyl)phenylphosphinic acid ≥98.0%

Supplier: TCI America

Synonyms: 3-(Hydroxy(phenyl)phosphoryl)propanoic acid, 3-Hydroxyphenylphosphinylpropanoic acid

C2887-25G C2887-5G
TCC2887-25GEA 113.76 CAD
TCC2887-25G TCC2887-5G
(2-Carboxyethyl)phenylphosphinic acid ≥98.0%
(2-Carboxyethyl)phenylphosphinic acid

CAS Number: 14657-64-8
MDL Number: MFCD08704562
Molecular Formula: C9H11O4P
Molecular Weight: 214.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 160

Formula: C₉H₁₁O₄P
MW: 214.16 g/mol
Melting Pt: 160 °C
MDL Number: MFCD08704562
CAS Number: 14657-64-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05b4717d0962d2b431a31eddf0c42d2b6f6ca1e0&vid=05b4717d0962d2b431a31eddf0c42d2b6f6ca1e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR