IR-813 p-Toluenesulfonate ≥98.0%
Supplier: TCI America
Synonyms:
2-(2-(2-Chloro-3-(2-(1,1,3-trimethyl-1H-benzo[e]indol-2(3H)-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-1,1,3-trimethyl-1H-benzo[e]indol-3-ium 4-methylbenzenesulfonate, IR 813 tosylate, 2-Chloro-3-[2-(1,1,3-trimethylbenz[e]indolin-2-ylidene)ethylidene]-1-[2-(1,1,3-t, 2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,1,3-trimethyl-2H-benz[e]indol-2-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-1,1,3-trimethyl-1H-benz[e]indolium 4-Methylbenzenesulphonate, 2-(2-(2-Chloro-3-(2-(1,1,3-trimethyl-1H-benzo[e]indol-2(3H)-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-1,1,3-trimethyl-1H-benzo[e]indol-3-ium 4-methylbenzenesulphonate, 2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,1,3-trimethyl-2H-benz[e]indol-2-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-1,1,3-trimethyl-1H-benz[e]indolium 4-Methylbenzenesulfonate
CAS Number: 134127-48-3
MDL Number: MFCD03093252
Molecular Formula: C47H47ClN2O3S
Molecular Weight: 755.41
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Lambda max.: 813 nm (MeOH)
Product Categories: Materials Chemistry, Near Infrared Absorption Materials
Formula:
C₄₇H₄₇ClN₂O₃S MW: 755.41 g/mol |
MDL Number:
MFCD03093252 CAS Number: 134127-48-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...