N-Carbobenzoxy-L-serine benzyl ester ≥97.0%

Supplier: TCI America

Synonyms: N-Cbz-L-serine Benzyl Ester, Z-Ser-OBzl, Benzyl ((benzyloxy)carbonyl)-L-serinate

C2873-25G C2873-5G
TCC2873-25GEA 423.44 CAD
TCC2873-25G TCC2873-5G
N-Carbobenzoxy-L-serine benzyl ester ≥97.0%
N-Carbobenzoxy-L-serine benzyl ester

CAS Number: 21209-51-8
MDL Number: MFCD00037824
Molecular Formula: C18H19NO5
Molecular Weight: 329.35
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 85
Specific rotation [a]20/D: 5 deg (C=1, CHCl3)


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Cbz-L-serine Benzyl Ester, Z-Ser-OBzl

Formula: C₁₈H₁₉NO₅
MW: 329.35 g/mol
Melting Pt: 85 °C
MDL Number: MFCD00037824
CAS Number: 21209-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a887f9dc70e0bed3af8bb5b4f9eb68e6be5e525d&vid=a887f9dc70e0bed3af8bb5b4f9eb68e6be5e525d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR