Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥97.0% (by GC)
Supplier: TCI America
C2872-1G
C2872-5G
TCC2872-1GEA
85.95
CAD
TCC2872-1G
TCC2872-5G
Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate ≥97.0% (by GC)
Benzyl-2,5-dihydro-1H-pyrrole-1-carboxylate
CAS Number: 31970-04-4
MDL Number: MFCD01863559
Molecular Formula: C12H13NO2
Molecular Weight: 203.24
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 110
Specific Gravity (20/20): 1.14
Other Product Names / Synonyms: N-Cbz-3-pyrroline
Formula:
C₁₂H₁₃NO₂ Boiling Pt: 126 °C Density: 1.14 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD01863559 CAS Number: 31970-04-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...