6-Chloro-3-formylchromone ≥98.0% (by GC)

Supplier: TCI America

C2869-25G C2869-5G
TCC2869-25GEA 222.78 CAD
TCC2869-25G TCC2869-5G
6-Chloro-3-formylchromone ≥98.0% (by GC)
6-Chloro-3-formylchromone

CAS Number: 42248-31-7
MDL Number: MFCD00139138
Molecular Formula: C10H5ClO3
Molecular Weight: 208.60
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 167


Other Product Names / Synonyms: 6-Chlorochromone-3-carboxaldehyde, 6-Chloro-4-oxo-4H-1-benzopyran-3-carboxaldehyde

Formula: C₁₀H₅ClO₃
Melting Pt: 167 °C
MDL Number: MFCD00139138
CAS Number: 42248-31-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4e6bb49fd08163718f861b04c02d426379ba5df&vid=b4e6bb49fd08163718f861b04c02d426379ba5df&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR