Clofazimine ≥98.0%

Supplier: TCI America

Synonyms: N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine, N,5-Bis(4-chlorophenyl)-3-(isopropylimino)-3,5-dihydrophenazin-2-amine

C2866-1G C2866-5G
TCC2866-1GEA 85.32 CAD
TCC2866-1G TCC2866-5G
Clofazimine ≥98.0%
Clofazimine

CAS Number: 2030-63-9
MDL Number: MFCD00056793
Molecular Formula: C27H22Cl2N4
Molecular Weight: 473.40
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 219


Product Categories: Bioscience, Synthetic Antimicrobial Ingredients, Antiprotozoal Ingredients
Other Product Names / Synonyms: N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine

Formula: C₂₇H₂₂Cl₂N₄
Melting Pt: 219 °C
MDL Number: MFCD00056793
CAS Number: 2030-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90fb361237f407a15e7b39fe4d69beb78eca6df5&vid=90fb361237f407a15e7b39fe4d69beb78eca6df5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR