Coumarin 153 ≥98.0% (by HPLC)

Supplier: TCI America

C2858-1G C2858-200MG
TCC2858-1GEA 767.88 CAD
TCC2858-1G TCC2858-200MG
Coumarin 153 ≥98.0% (by HPLC)
Coumarin 153

CAS Number: 53518-18-6
MDL Number: MFCD00041843
Molecular Formula: C16H14F3NO2
Molecular Weight: 309.29
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 166
Lambda max.: 426 nm (H2O)


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Coumarin Dyes (Functional Organic Dyes), Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Coumarin 540A, 2,3,6,7-Tetrahydro-9-(trifluoromethyl)-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one

Formula: C₁₆H₁₄F₃NO₂
Melting Pt: 166 °C
MDL Number: MFCD00041843
CAS Number: 53518-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9b626727a2b50031d1e5377940afda5fa44c31b&vid=e9b626727a2b50031d1e5377940afda5fa44c31b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR