Cedrol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
(+)-Cedrol, (3R,3aS,6R,7R,8aS)-3,6,8,8-Tetramethyloctahydro-1H-3a,7-methanoazulen-6-ol
C2854-25G
C2854-5G
TCC2854-25GEA
391.84
CAD
TCC2854-25G
TCC2854-5G
Cedrol ≥98.0% (by GC)
Cedrol
CAS Number: 77-53-2
MDL Number: MFCD00062952
Molecular Formula: C15H26O
Molecular Weight: 222.37
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 273
Melting point (°C): 86
Specific rotation [a]20/D: 10 deg (C=5, CHCl3)
Formula:
C₁₅H₂₆O MW: 222.37 g/mol Boiling Pt: 273 °C Melting Pt: 86 °C |
MDL Number:
MFCD00062952 CAS Number: 77-53-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...