2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid ≥97.0% (by HPLC)

Supplier: TCI America

C2848-25G C2848-5G
TCC2848-25GEA 184.86 CAD
TCC2848-25G TCC2848-5G
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid ≥97.0% (by HPLC)
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid

CAS Number: 39608-31-6
MDL Number: MFCD00021747
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >97.0% (HPLC)
Form: Clear Liquid


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-N-methylglycine, Z-N-Me-Gly-OH

Formula: C₁₁H₁₃NO₄
MDL Number: MFCD00021747
CAS Number: 39608-31-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bfd7b524fb817b6c2411989595d0e26dc399e1b&vid=4bfd7b524fb817b6c2411989595d0e26dc399e1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR