1,2,3,4-Cyclobutanetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis), purified by sublimation
Supplier: TCI America
Synonyms:
Cyclobuta[1,2-c:3,4-c']difuran-1,3,4,6(3aH,3bH,6aH,6bH)-tetraone
C2842-1G
C2842-5G
TCC2842-1GEA
58.46
CAD
TCC2842-1G
TCC2842-5G
1,2,3,4-Cyclobutanetetracarboxylic dianhydride ≥98.0% (by titrimetric analysis), purified by sublimation
1,2,3,4-Cyclobutanetetracarboxylic dianhydride
CAS Number: 4415-87-6
MDL Number: MFCD00004944
Molecular Formula: C8H4O6
Molecular Weight: 196.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 285
Formula:
C₈H₄O₆ MW: 196.11 g/mol Melting Pt: 285 °C |
MDL Number:
MFCD00004944 CAS Number: 4415-87-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...