7-Chloroisatin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C2840-1G C2840-5G
TCC2840-1GEA 45.82 CAD
TCC2840-1G TCC2840-5G
7-Chloroisatin ≥98.0% (by GC, titration analysis)
7-Chloroisatin

CAS Number: 7477-63-6
MDL Number: MFCD00022796
Molecular Formula: C8H4ClNO2
Molecular Weight: 181.58
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 190
Lambda max.: 404 nm (EtOH)


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 7-Chloroindole-2,3-dione, 7-Chloro-2,3-indolinedione

Formula: C₈H₄ClNO₂
Melting Pt: 190 °C
MDL Number: MFCD00022796
CAS Number: 7477-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=165d9cdecb9262e09a1b4fe94eae62ac9eb1245b&vid=165d9cdecb9262e09a1b4fe94eae62ac9eb1245b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR