N-Carbobenzoxy-D-leucine ≥97.0%

Supplier: TCI America

Synonyms: Z-D-Leu-OH, N-Cbz-D-leucine

C2839-25G C2839-5G
TCC2839-25GEA 660.44 CAD
TCC2839-25G TCC2839-5G
N-Carbobenzoxy-D-leucine ≥97.0%
N-Carbobenzoxy-D-leucine

CAS Number: 28862-79-5
MDL Number: MFCD00066068
Molecular Formula: C14H19NO4
Molecular Weight: 265.31
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Clear Liquid
Specific rotation [a]20/D: 17 deg (C=5, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Cbz-D-leucine, Z-D-Leu-OH

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD00066068
CAS Number: 28862-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54a0238ce6f0250d49b28eceadfad0b8b38485d&vid=f54a0238ce6f0250d49b28eceadfad0b8b38485d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR