N-Carbobenzoxyglycine methyl ester ≥95.0%
Supplier: TCI America
Synonyms:
N-Cbz-glycine Methyl Ester, N-Benzyloxycarbonylglycine methyl ester, Z-Gly-OMe
C2838-25G
C2838-5G
TCC2838-25GEA
134.3
CAD
TCC2838-25G
TCC2838-5G
N-Carbobenzoxyglycine methyl ester ≥95.0%
N-Carbobenzoxyglycine methyl ester
CAS Number: 1212-53-9
MDL Number: MFCD00043631
Molecular Formula: C11H13NO4
Molecular Weight: 223.23
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Specific Gravity (20/20): 1.19
Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Cbz-glycine Methyl Ester, Z-Gly-OMe
Formula:
C₁₁H₁₃NO₄ MW: 223.23 g/mol Boiling Pt: 151 °C Density: 1.19 (20 °C) |
MDL Number:
MFCD00043631 CAS Number: 1212-53-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...