7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Coumarin 30, 3-(2-N-Methylbenzimidazolyl)-7-N,N-diethylaminocoumarin, 7-(Diethylamino)-3-(1-methyl-1H-benzo[d]imidazol-2-yl)-2H-chromen-2-one

C2837-1G C2837-200MG
TCC2837-1GEA 349.18 CAD
TCC2837-1G TCC2837-200MG
7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin ≥98.0% (by HPLC, titration analysis)
7-(Diethylamino)-3-(1-methyl-2-benzimidazolyl)coumarin

CAS Number: 41044-12-6
MDL Number: MFCD00051349
Molecular Formula: C21H21N3O2
Molecular Weight: 347.42
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 223
Lambda max.: 408 nm (CH3CN)


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Coumarin Dyes (Functional Organic Dyes), Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: Coumarin 30, 3-(2-N-Methylbenzimidazolyl)-7-N,N-diethylaminocoumarin

Formula: C₂₁H₂₁N₃O₂
MW: 347.42 g/mol
Melting Pt: 223 °C
MDL Number: MFCD00051349
CAS Number: 41044-12-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=001b211cebf6c15987456fb80cdb9d8d1a9fa3dc&vid=001b211cebf6c15987456fb80cdb9d8d1a9fa3dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR