4-Chloro-2-methylpyrimidine ≥98.0% (by GC)
Supplier: TCI America
C2835-1G
C2835-200MG
TCC2835-1GEA
543.84
CAD
TCC2835-1G
TCC2835-200MG
4-Chloro-2-methylpyrimidine ≥98.0% (by GC)
4-Chloro-2-methylpyrimidine
CAS Number: 4994-86-9
MDL Number: MFCD00234069
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 168
Melting point (°C): 57
Formula:
C₅H₅ClN₂ Boiling Pt: 168 °C Melting Pt: 57 °C |
MDL Number:
MFCD00234069 CAS Number: 4994-86-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...