3-Cyanoumbelliferone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-Cyano-7-hydroxycoumarin, 7-Hydroxy-2-oxo-2H-chromene-3-carbonitrile

C2834-1G C2834-5G
TCC2834-1GEA 139.04 CAD
TCC2834-1G TCC2834-5G
3-Cyanoumbelliferone ≥98.0% (by GC)
3-Cyanoumbelliferone

CAS Number: 19088-73-4
MDL Number: MFCD00037480
Molecular Formula: C10H5NO3
Molecular Weight: 187.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 280
Lambda max.: 450 nm (MeOH)


Other Product Names / Synonyms: 3-Cyano-7-hydroxycoumarin

Formula: C₁₀H₅NO₃
MW: 187.15 g/mol
Melting Pt: 280 °C
MDL Number: MFCD00037480
CAS Number: 19088-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52638122838b3094c7011ca8e074ba89f8d6290e&vid=52638122838b3094c7011ca8e074ba89f8d6290e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR