(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2832-1G C2832-5G
TCC2832-1GEA 403.37 CAD
TCC2832-1G TCC2832-5G
(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide ≥98.0% (by HPLC, titration analysis)
(S)-6-Chloro-4-hydroxy-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-1,1-dioxide

CAS Number: 160982-16-1
MDL Number: MFCD09033311
Molecular Formula: C6H6ClNO3S2
Molecular Weight: 239.69
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 130
Specific rotation [a]20/D: -5 deg (C=1, MeOH)

Formula: C₆H₆ClNO₃S₂
Melting Pt: 130 °C
MDL Number: MFCD09033311
CAS Number: 160982-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=027f84ae23dbf363ffff3d1d475a21e20c1cac56&vid=027f84ae23dbf363ffff3d1d475a21e20c1cac56&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR