3-Cyanocoumarin ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Coumarin-3-carbonitrile, 2-Oxo-2H-chromene-3-carbonitrile

C2830-1G C2830-5G
TCC2830-1GEA 36.34 CAD
TCC2830-1G TCC2830-5G
3-Cyanocoumarin ≥98.0% (by GC)
3-Cyanocoumarin

CAS Number: 15119-34-3
MDL Number: MFCD00115699
Molecular Formula: C10H5NO2
Molecular Weight: 171.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 186
Lambda max.: 358 nm (DMSO)


Other Product Names / Synonyms: 3-Cyanocoumarin

Formula: C₁₀H₅NO₂
MW: 171.16 g/mol
Melting Pt: 186 °C
MDL Number: MFCD00115699
CAS Number: 15119-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d16397fb256ce8d3961638d15d1a198dcc00dac0&vid=d16397fb256ce8d3961638d15d1a198dcc00dac0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR