4-Chloro-2,3-dimethylpyridine-N-oxide ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Chloro-2,3-dimethylpyridine 1-oxide

C2820-25G C2820-5G
TCC2820-25GEA 246.32 CAD
TCC2820-25G TCC2820-5G
4-Chloro-2,3-dimethylpyridine-N-oxide ≥98.0% (by GC, titration analysis)
4-Chloro-2,3-dimethylpyridine-N-oxide

CAS Number: 59886-90-7
MDL Number: MFCD06411125
Molecular Formula: C7H8ClNO
Molecular Weight: 157.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 108
Storage Temperature: <0°C

Formula: C₇H₈ClNO
MW: 157.60 g/mol
Melting Pt: 108 °C
MDL Number: MFCD06411125
CAS Number: 59886-90-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72708be96962d91c034b6a42312b70b38b7f9859&vid=72708be96962d91c034b6a42312b70b38b7f9859&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR