ɑ-Cyclohexylphenylacetic acid ≥95.0% (by GC, titration analysis)
Supplier: TCI America
C2812-1G
C2812-5G
TCC2812-1GEA
34.76
CAD
TCC2812-1G
TCC2812-5G
ɑ-Cyclohexylphenylacetic acid ≥95.0% (by GC, titration analysis)
alpha-Cyclohexylphenylacetic acid
CAS Number: 3894-09-5
MDL Number: MFCD00001477
Molecular Formula: C14H18O2
Molecular Weight: 218.30
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Melting point (°C): 152
Other Product Names / Synonyms: 2-Cyclohexyl-2-phenylacetic Acid
Formula:
C₁₄H₁₈O₂ Melting Pt: 152 °C |
MDL Number:
MFCD00001477 CAS Number: 3894-09-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...