5-(2-Chlorophenyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2811-1G C2811-5G
TCC2811-1GEA 52.14 CAD
TCC2811-1G TCC2811-5G
5-(2-Chlorophenyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-(2-Chlorophenyl)-1H-tetrazole

CAS Number: 50907-46-5
MDL Number: MFCD00040950
Molecular Formula: C7H5ClN4
Molecular Weight: 180.60
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 179
Lambda max.: 234 nm

Formula: C₇H₅ClN₄
Melting Pt: 179 °C
MDL Number: MFCD00040950
CAS Number: 50907-46-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b51900c4eeb96a8b8e930cdf953b3ecf4258157&vid=5b51900c4eeb96a8b8e930cdf953b3ecf4258157&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR