Cesium green ≥85.0%

Supplier: TCI America

Synonyms: 4''-[2-[2-[2-[2-(4-Nitrophenoxy)ethoxy]ethoxy]ethoxy]ethoxy]-[1,1':4',1''-terphenyl]-4-ol

C2806-250MG C2806-50MG
TCC2806-250MGEA 920.67 CAD
TCC2806-250MG TCC2806-50MG
Cesium green ≥85.0%
Cesium green

CAS Number: 1611477-45-2
Molecular Formula: C32H33NO8
Molecular Weight: 559.62
Purity/Analysis Method: >85.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 4''-[2-[2-[2-[2-(4-Nitrophenoxy)ethoxy]ethoxy]ethoxy]ethoxy]-[1,1':4',1''-terphenyl]-4-ol

Formula: C₃₂H₃₃NO₈
CAS Number: 1611477-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cba7bd0ab0ceced8a8bb0938b1002cb7285185e0&vid=cba7bd0ab0ceced8a8bb0938b1002cb7285185e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR