1-Carbobenzoxy-3-piperidinecarboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2805-1G C2805-5G
TCC2805-1GEA 56.25 CAD
TCC2805-1G TCC2805-5G
1-Carbobenzoxy-3-piperidinecarboxylic acid ≥97.0% (by GC, titration analysis)
1-Carbobenzoxy-3-piperidinecarboxylic acid

CAS Number: 78190-11-1
MDL Number: MFCD02931282
Molecular Formula: C14H17NO4
Molecular Weight: 263.29
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 97


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 1-Cbz-3-piperidinecarboxylic Acid, 1-Carbobenzoxynipecotic Acid, 1-Cbz-nipecotic Acid

Formula: C₁₄H₁₇NO₄
Melting Pt: 97 °C
MDL Number: MFCD02931282
CAS Number: 78190-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=661cd0936c8eedc89d32b1f8c58173585ee9f4fd&vid=661cd0936c8eedc89d32b1f8c58173585ee9f4fd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR