4-Chloro-N-methylpicolinamide ≥98.0%

Supplier: TCI America

Synonyms: 4-Chloro-N-methylpyridine-2-carboxamide

C2793-1G C2793-5G
TCC2793-1GEA 54.51 CAD
TCC2793-1G TCC2793-5G
4-Chloro-N-methylpicolinamide ≥98.0%
4-Chloro-N-methylpicolinamide

CAS Number: 220000-87-3
MDL Number: MFCD02185921
Molecular Formula: C7H7ClN2O
Molecular Weight: 170.60
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 47


Other Product Names / Synonyms: 4-Chloro-N-methylpicolinamide

Formula: C₇H₇ClN₂O
MW: 170.60 g/mol
Melting Pt: 47 °C
MDL Number: MFCD02185921
CAS Number: 220000-87-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5692917d3b75e62970ae832008a08ae91a3a52f9&vid=5692917d3b75e62970ae832008a08ae91a3a52f9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR