2-Chloro-6-methylpyrazine ≥98.0% (by GC)

Supplier: TCI America

C2777-1G C2777-5G
TCC2777-1GEA 172.22 CAD
TCC2777-1G TCC2777-5G
2-Chloro-6-methylpyrazine ≥98.0% (by GC)
2-Chloro-6-methylpyrazine

CAS Number: 38557-71-0
MDL Number: MFCD00055032
Molecular Formula: C5H5ClN2
Molecular Weight: 128.56
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 83
Melting point (°C): 52

Formula: C₅H₅ClN₂
MW: 128.5596 g/mol
Boiling Pt: 83 °C
Melting Pt: 52 °C
MDL Number: MFCD00055032
CAS Number: 38557-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c488945dc00cb2c74692583421944cff09b2a1c0&vid=c488945dc00cb2c74692583421944cff09b2a1c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR