6-Chloro-o-toluic acid ≥98.0%
Supplier: TCI America
Synonyms:
2-Chloro-6-methylbenzoic acid
C2776-1G
C2776-5G
TCC2776-1GEA
135.88
CAD
TCC2776-1G
TCC2776-5G
6-Chloro-o-toluic acid ≥98.0%
6-Chloro-o-toluic acid
CAS Number: 21327-86-6
MDL Number: MFCD00045799
Molecular Formula: C8H7ClO2
Molecular Weight: 170.59
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 106
Other Product Names / Synonyms: 6-Chloro-o-toluic Acid
Formula:
C₈H₇ClO₂ Melting Pt: 106 °C |
MDL Number:
MFCD00045799 CAS Number: 21327-86-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...