N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2773-25G C2773-5G
TCC2773-25GEA 766.93 CAD
TCC2773-25G TCC2773-5G
N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine ≥98.0% (by HPLC, titration analysis)
N-Carbobenzoxy-4-hydroxy-D-2-phenylglycine

CAS Number: 26787-75-7
Molecular Formula: C16H15NO5
Molecular Weight: 301.30
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -127 deg (C=10, MeOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: (R)-2-(Carbobenzoxy)amino-2-(4-hydroxyphenyl)acetic Acid, Z-D-Phg(4-OH)-OH

Formula: C₁₆H₁₅NO₅
MDL Number: MFCD09756650
CAS Number: 26787-75-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a50db7eeb6109b20cd65bf2d79c921bf8684f4cc&vid=a50db7eeb6109b20cd65bf2d79c921bf8684f4cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR