2-((4-Cyanophenyl)amino)acetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-(4-CN-Phenyl)-Gly-OH, N-(4-Cyanophenyl)glycine

C2771-25G C2771-5G
TCC2771-25GEA 143.78 CAD
TCC2771-25G TCC2771-5G
2-((4-Cyanophenyl)amino)acetic acid ≥98.0% (by HPLC, titration analysis)
2-((4-Cyanophenyl)amino)acetic acid

CAS Number: 42288-26-6
MDL Number: MFCD01464022
Molecular Formula: C9H8N2O2
Molecular Weight: 176.18
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 237

Formula: C₉H₈N₂O₂
MW: 176.1700 g/mol
Melting Pt: 237 °C
MDL Number: MFCD01464022
CAS Number: 42288-26-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5846b304f25dcc884900170fa3e37b012a4a1bf&vid=e5846b304f25dcc884900170fa3e37b012a4a1bf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR