2-(1-Methylguanidino)ethyl dihydrogen phosphate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

C2767-25G C2767-5G
TCC2767-25GEA 61.62 CAD
TCC2767-25G TCC2767-5G
2-(1-Methylguanidino)ethyl dihydrogen phosphate ≥98.0% (by HPLC, titration analysis)
2-(1-Methylguanidino)ethyl dihydrogen phosphate

CAS Number: 6903-79-3
MDL Number: MFCD00868297
Molecular Formula: C4H12N3O4P
Molecular Weight: 197.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 244
Melting point (°C): 244


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 2-(1-Methylguanidino)ethyl Dihydrogen Phosphate

Formula: C₄H₁₂N₃O₄P
Boiling Pt: 244 °C
Melting Pt: 244 °C
MDL Number: MFCD00868297
CAS Number: 6903-79-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2cee3d1e7d331dccc84cf3ebfd4d5593bacfeae&vid=c2cee3d1e7d331dccc84cf3ebfd4d5593bacfeae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR