4-Cyano-4'-propylbiphenyl ≥99.0% (by GC)

Supplier: TCI America

C2764-1G C2764-5G
TCC2764-1GEA 45.82 CAD
TCC2764-1G TCC2764-5G
4-Cyano-4'-propylbiphenyl ≥99.0% (by GC)
4-Cyano-4'-propylbiphenyl

CAS Number: 58743-76-3
MDL Number: MFCD00505987
Molecular Formula: C16H15N
Molecular Weight: 221.30
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 67


Other Product Names / Synonyms: 4'-Propyl-4-biphenylcarbonitrile, 3CB

Formula: C₁₆H₁₅N
Melting Pt: 67 °C
MDL Number: MFCD00505987
CAS Number: 58743-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8fb4de1c355baffc882552724a802fdb2c5dcd78&vid=8fb4de1c355baffc882552724a802fdb2c5dcd78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR